Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp CORTECS C8 Method Validation Kit, 1.6 μm, 3x50 mm

Catalog No. 502391877 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

CORTECS C8 method validation kits provide columns of three different batches. CORTECS C8 columns are used for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 phases.

  • C8 chemistry.
  • Reversed phase separation.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Max pressure: 18000 psi (1240 bar).
  • Endcapped.
  • Spherical particle shape.
  • Particle size: 1.6 μm.
  • Parker-style endfitting.
  • Pore size: 90 Å.
  • Method validation kit format.
  • UPLC, UHPLC system compatible.
  • Solid Core particle technology.
  • USP classification L7.
  • Dimensions: 3 mm x 50 mm.
  • Carbon load: 4.5%.
  • eCord included.
  • 3 units per package.

Catalog No. 50-239-1877 Supplier Waters Corp Supplier No. 186008414
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CORTECS C8 method validation kits provide columns of three different batches. CORTECS C8 columns are used for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 phases.

  • C8 chemistry.
  • Reversed phase separation.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Max pressure: 18000 psi (1240 bar).
  • Endcapped.
  • Spherical particle shape.
  • Particle size: 1.6 μm.
  • Parker-style endfitting.
  • Pore size: 90 Å.
  • Method validation kit format.
  • UPLC, UHPLC system compatible.
  • Solid Core particle technology.
  • USP classification L7.
  • Dimensions: 3 mm x 50 mm.
  • Carbon load: 4.5%.
  • eCord included.
  • 3 units per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.