Learn More
Waters Corp CORTECS C8 Method Validation Kit, 90 Å, 2.7 μm, 4.6 x 50 mm
Catalog No. 502391951 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit is designed for rigorous method validation applications. It features a CORTECS C8 phase, optimized for efficient chromatographic separations.
  • 90 Å pore size.
  • 2.7 μm particle size.
  • Kit contains 3 columns.
  • Column dimensions are 4.6 x 50 mm.
  • Complies with USP L7 classification.
Catalog No. 50-239-1951 Supplier Waters Corp Supplier No. 186008374
This Method Validation Kit is designed for rigorous method validation applications. It features a CORTECS C8 phase, optimized for efficient chromatographic separations.
  • 90 Å pore size.
  • 2.7 μm particle size.
  • Kit contains 3 columns.
  • Column dimensions are 4.6 x 50 mm.
  • Complies with USP L7 classification.