Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp CORTECS C8 Kit, 90Å, 1.6 μm, 3 mm X 75 mm

Catalog No. 502392166 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This CORTECS C8 method validation kit includes columns from three different batches, designed for general-purpose method development under low pH test conditions. Its C8 ligand offers lower hydrophobicity and retention compared to C18 phases, making it suitable for various assays.

  • Includes three columns from different batches for method validation.
  • Utilized for general-purpose method development under low pH test conditions.
  • C8 ligand offers lower hydrophobicity and retention than C18 phases.
  • Suitable for UPLC and UHPLC systems.
  • Pore size: 90 Å.
  • Particle size: 1.6 μm.
  • Dimensions: 3 mm × 75 mm.

Catalog No. 50-239-2166 Supplier Waters Corp Supplier No. 186008415
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This CORTECS C8 method validation kit includes columns from three different batches, designed for general-purpose method development under low pH test conditions. Its C8 ligand offers lower hydrophobicity and retention compared to C18 phases, making it suitable for various assays.

  • Includes three columns from different batches for method validation.
  • Utilized for general-purpose method development under low pH test conditions.
  • C8 ligand offers lower hydrophobicity and retention than C18 phases.
  • Suitable for UPLC and UHPLC systems.
  • Pore size: 90 Å.
  • Particle size: 1.6 μm.
  • Dimensions: 3 mm × 75 mm.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.