Learn More
Waters Corp UPLC Columns Method Validation Kit, C8, 2.7 μm, 2.1 × 50 mm
Catalog No. 502391633 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit evaluates the quality, reliability, and consistency of a method by including three batches of chromatographic media, derived from different base particles. Each kit contains three columns from three batches of material.
  • Chemistry: C8
  • Particle size: 2.7 μm
  • Dimension: 2.1 × 50 mm
Catalog No. 50-239-1633 Supplier Waters Corp Supplier No. 186008354
This Method Validation Kit evaluates the quality, reliability, and consistency of a method by including three batches of chromatographic media, derived from different base particles. Each kit contains three columns from three batches of material.
  • Chemistry: C8
  • Particle size: 2.7 μm
  • Dimension: 2.1 × 50 mm