Learn More
Waters Corp HPLC Column, Silica C8, 75 x 4.6 mm, 2.7 μm
Catalog No. 502392044 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC Column CORTECS Method Validation Kit is a Silica C8 column designed for analytical applications. It features high-value analytical technologies, suitable for method validation processes in chromatography, mass spectrometry, and thermal analysis.
  • Designed for method validation.
  • Made from Silica C8 material.
  • Dimensions of 75 mm length x 4.6 mm diameter.
  • Features a particle size of 2.7 μm.
  • Has a pore size of 90 Å.
  • Packaged with 3 columns per pack.
Catalog No. 50-239-2044 Supplier Waters Corp Supplier No. 186008375
This HPLC Column CORTECS Method Validation Kit is a Silica C8 column designed for analytical applications. It features high-value analytical technologies, suitable for method validation processes in chromatography, mass spectrometry, and thermal analysis.
  • Designed for method validation.
  • Made from Silica C8 material.
  • Dimensions of 75 mm length x 4.6 mm diameter.
  • Features a particle size of 2.7 μm.
  • Has a pore size of 90 Å.
  • Packaged with 3 columns per pack.