Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp CORTECS C8 Method Validation Kit, 1.6 μm, 3 mm x 100 mm, 90 Å, 3 pk

Catalog No. 502391257 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The CORTECS C8 method validation kits provide three columns of different batches. These columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and thus less retentive compared to C18 phases, making it suitable for specific assays.

  • Chemistry: C8
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2 pH to 8 pH
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 1.6 μm
  • Pore size: 90 Å
  • Inner diameter: 3 mm
  • Length: 100 mm

Catalog No. 50-239-1257 Supplier Waters Corp Supplier No. 186008416
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The CORTECS C8 method validation kits provide three columns of different batches. These columns are designed for general purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and thus less retentive compared to C18 phases, making it suitable for specific assays.

  • Chemistry: C8
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2 pH to 8 pH
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 1.6 μm
  • Pore size: 90 Å
  • Inner diameter: 3 mm
  • Length: 100 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.