Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp CORTECS C8 Method Validation Kit, 90Å, 2.7 μm

Catalog No. 502390943 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This CORTECS C8 Method Validation Kit is designed for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 phases, making it suitable for method validation purposes.

  • Particle size: 2.7 μm
  • Pore size: 90 Å
  • Dimensions: 3 mm x 75 mm
  • Quantity per package: 3 pk
  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45 °C
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle technology: Solid core
  • Format: Column
  • System: UPLC, UHPLC
  • Surface area: 100
  • Endfitting type: Parker-style
  • Particle shape: Spherical
  • Silanol activity: Low
  • Endcapped: Yes

Catalog No. 50-239-0943 Supplier Waters Corp Supplier No. 186008365
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This CORTECS C8 Method Validation Kit is designed for general purpose method development under low pH test conditions. The C8 ligand is preferred for some assays because it is less hydrophobic and therefore less retentive compared to C18 phases, making it suitable for method validation purposes.

  • Particle size: 2.7 μm
  • Pore size: 90 Å
  • Dimensions: 3 mm x 75 mm
  • Quantity per package: 3 pk
  • Chemistry: C8
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45 °C
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle technology: Solid core
  • Format: Column
  • System: UPLC, UHPLC
  • Surface area: 100
  • Endfitting type: Parker-style
  • Particle shape: Spherical
  • Silanol activity: Low
  • Endcapped: Yes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.