Learn More
Waters Corp VanGuard Cartridge, C8, 90 Å Pore, 2.7 μm Particle
Catalog No. 502392160 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C8 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by filtering out particulate contamination from the mobile phase stream. This specific cartridge is optimized for protecting all 4.6 mm ID CORTECS C8 analytical columns that utilize 2.7 μm sorbent particles.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 1240 bar (18000 psi)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 2.7 μm particle size
  • Parker-style endfitting type
  • 90 Å pore size
  • 100 surface area
  • Compatible with UPLC and UHPLC systems
  • Solid core particle technology
  • L7 USP classification
  • 3.9 mm inner diameter
  • 5 mm length
  • 4.5 % carbon load
  • Features eCord technology
Catalog No. 50-239-2160 Supplier Waters Corp Supplier No. 186008422
CORTECS C8 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by filtering out particulate contamination from the mobile phase stream. This specific cartridge is optimized for protecting all 4.6 mm ID CORTECS C8 analytical columns that utilize 2.7 μm sorbent particles.
  • C8 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 1240 bar (18000 psi)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 2.7 μm particle size
  • Parker-style endfitting type
  • 90 Å pore size
  • 100 surface area
  • Compatible with UPLC and UHPLC systems
  • Solid core particle technology
  • L7 USP classification
  • 3.9 mm inner diameter
  • 5 mm length
  • 4.5 % carbon load
  • Features eCord technology