Learn More
Waters Corp CORTECS Column, C18, 90Å, 2.7 μm, 3 mm × 100 mm
Catalog No. NC3908139 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases, and neutrals at low and mid-range pH.
  • Solid-core particle technology for increased efficiency, performance, and higher throughput.
  • Increased analyte loading capacity with faster mass transfer kinetics for uniform packing.
  • Utilizes ultra-low dispersion column hardware and optimal packing for efficient separation.
  • Operates within a pH range of 2 to 8, max temperature of 45 °C, and 18000 psi.
  • Features low silanol activity and a 6.6% carbon load for excellent resolution and selectivity.
  • Ensures excellent peak shape and batch-to-batch reproducibility for various compounds.
  • Includes a Certificate of Analysis and Performance Test Chromatogram.
  • Compatible with VanGuard Cartridges to extend analytical column lifetime.
Catalog No. NC3908139 Supplier Waters Corp Supplier No. 186007372
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases, and neutrals at low and mid-range pH.
  • Solid-core particle technology for increased efficiency, performance, and higher throughput.
  • Increased analyte loading capacity with faster mass transfer kinetics for uniform packing.
  • Utilizes ultra-low dispersion column hardware and optimal packing for efficient separation.
  • Operates within a pH range of 2 to 8, max temperature of 45 °C, and 18000 psi.
  • Features low silanol activity and a 6.6% carbon load for excellent resolution and selectivity.
  • Ensures excellent peak shape and batch-to-batch reproducibility for various compounds.
  • Includes a Certificate of Analysis and Performance Test Chromatogram.
  • Compatible with VanGuard Cartridges to extend analytical column lifetime.