Learn More
Waters Corp CORTECS HILIC Column, 90Å, 1.6 μm, 2.1 mm x 30 mm
Catalog No. 502392964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle. They are designed to retain extremely polar analytes and offer orthogonal selectivity versus C18 columns.
  • High-efficiency columns based on a solid-core particle
  • Designed to retain extremely polar analytes
  • Offer orthogonal selectivity versus C18 columns
  • eCord enabled
Catalog No. 50-239-2964 Supplier Waters Corp Supplier No. 176003156
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle. They are designed to retain extremely polar analytes and offer orthogonal selectivity versus C18 columns.
  • High-efficiency columns based on a solid-core particle
  • Designed to retain extremely polar analytes
  • Offer orthogonal selectivity versus C18 columns
  • eCord enabled