Learn More
Waters Corp Waters CORTECS UPLC Column, HILIC, 2.7 μm, 2.1 × 75 mm
Catalog No. 502391429 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS UPLC Columns are designed as the performance standard, featuring sub-2-μm, solid-core particle technology for high column efficiencies with low-dispersive UPLC instrumentation. This specific HILIC chemistry column offers flexibility for rapidly separating a wide array of compounds.
  • Features sub-2-μm, solid-core particle technology
  • Provides high column efficiencies with low-dispersive UPLC instrumentation
  • Produces sharper, narrower peaks compared to fully porous particles of similar size
  • Ideal for increased resolution, speed, and sensitivity
Catalog No. 50-239-1429 Supplier Waters Corp Supplier No. 186007381
CORTECS UPLC Columns are designed as the performance standard, featuring sub-2-μm, solid-core particle technology for high column efficiencies with low-dispersive UPLC instrumentation. This specific HILIC chemistry column offers flexibility for rapidly separating a wide array of compounds.
  • Features sub-2-μm, solid-core particle technology
  • Provides high column efficiencies with low-dispersive UPLC instrumentation
  • Produces sharper, narrower peaks compared to fully porous particles of similar size
  • Ideal for increased resolution, speed, and sensitivity