Learn More
Waters Corp Waters Corporation CORTECS HILIC Column, 2.7 μm, 3.0 x 75 mm, 3-Pack
Catalog No. 502391570 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS HILIC 2.7 μm columns are designed for retaining very polar compounds using hydrophilic-interaction chromatography (HILIC). This orthogonal separation mode to reversed-phase chromatography benefits from acetonitrile-rich mobile phases and an unbonded HILIC stationary phase, offering increased mass spectrometry response and direct injection of organic extracts (e.g., SPE, LLE, PPT).
  • Superior resolution and retention of polar analytes.
  • Orthogonal selectivity versus C18 columns.
  • Increased mass spectrometry response.
  • Direct injection for organic extracts.
  • Maximizes HPLC/UHPLC efficiency for enhanced resolution.
  • Excellent batch-to-batch reproducibility.
  • Resistance to bonded phase loss for good column lifetimes.
  • Lower operating backpressure without sacrificing efficiency.
  • Simple method transfer across HPLC, UHPLC, and UPLC systems.
Catalog No. 50-239-1570 Supplier Waters Corp Supplier No. 176003286
Add to Cart
Add to Cart
CORTECS HILIC 2.7 μm columns are designed for retaining very polar compounds using hydrophilic-interaction chromatography (HILIC). This orthogonal separation mode to reversed-phase chromatography benefits from acetonitrile-rich mobile phases and an unbonded HILIC stationary phase, offering increased mass spectrometry response and direct injection of organic extracts (e.g., SPE, LLE, PPT).
  • Superior resolution and retention of polar analytes.
  • Orthogonal selectivity versus C18 columns.
  • Increased mass spectrometry response.
  • Direct injection for organic extracts.
  • Maximizes HPLC/UHPLC efficiency for enhanced resolution.
  • Excellent batch-to-batch reproducibility.
  • Resistance to bonded phase loss for good column lifetimes.
  • Lower operating backpressure without sacrificing efficiency.
  • Simple method transfer across HPLC, UHPLC, and UPLC systems.