Learn More
Waters Corp CORTECS HILIC Column, 90Å, 2.7 μm, 3 mm x 150 mm
Catalog No. 502392161 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS HILIC Columns offer high-efficiency separation for extremely polar analytes using solid-core particles. They provide orthogonal selectivity to C18 columns, allowing resolution of complex mixtures with high performance or increased throughput.
  • Enhanced performance from solid-core particles, reducing van Deemter equation terms for efficient separations.
  • Designed for Hydrophilic Interaction Chromatography (HILIC), ideal for retaining polar compounds not suitable for reversed-phase.
  • Maintains high stability and retention.
  • Uses mobile phases with high organic solvent concentration, beneficial for polar analytes and quicker sample preparation.
  • Facilitates direct interaction with SPE eluates, increasing sample throughput.
  • Manufactured to high standards for consistent quality and reliability.
Catalog No. 50-239-2161 Supplier Waters Corp Supplier No. 186007388
CORTECS HILIC Columns offer high-efficiency separation for extremely polar analytes using solid-core particles. They provide orthogonal selectivity to C18 columns, allowing resolution of complex mixtures with high performance or increased throughput.
  • Enhanced performance from solid-core particles, reducing van Deemter equation terms for efficient separations.
  • Designed for Hydrophilic Interaction Chromatography (HILIC), ideal for retaining polar compounds not suitable for reversed-phase.
  • Maintains high stability and retention.
  • Uses mobile phases with high organic solvent concentration, beneficial for polar analytes and quicker sample preparation.
  • Facilitates direct interaction with SPE eluates, increasing sample throughput.
  • Manufactured to high standards for consistent quality and reliability.