Learn More
Waters Corp CORTECS HILIC Column, 90Å, 2.7 μm, 3 mm × 30 mm
Catalog No. 502392052 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle that are designed to retain extremely polar analytes. They offer orthogonal selectivity versus C18 columns.
  • High-efficiency columns for retaining extremely polar analytes.
  • Offers orthogonal selectivity versus C18 columns.
Catalog No. 50-239-2052 Supplier Waters Corp Supplier No. 176003284
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle that are designed to retain extremely polar analytes. They offer orthogonal selectivity versus C18 columns.
  • High-efficiency columns for retaining extremely polar analytes.
  • Offers orthogonal selectivity versus C18 columns.