Learn More
Waters Corp HILIC Column, 90Å, 2.7 μm, 150 mm
Catalog No. 502390599 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns that utilize Hydrophilic Interaction Chromatography (HILIC) mode. They are designed with solid-core particles to retain extremely polar analytes and offer orthogonal selectivity compared to C18 columns.
  • Achieves excellent efficiency and performance
  • Resolves complicated mixtures and increases throughput
  • High level of retention and stability
  • Enables faster sample preparation with high organic solvent concentration mobile phases
  • Increases MS response and sensitivity
  • Reduces A, B, and C terms of the van Deemter equation for improved performance
  • Manufactured to high standards under strict and controlled environments
Catalog No. 50-239-0599 Supplier Waters Corp Supplier No. 186007383
CORTECS HILIC Columns are high-efficiency columns that utilize Hydrophilic Interaction Chromatography (HILIC) mode. They are designed with solid-core particles to retain extremely polar analytes and offer orthogonal selectivity compared to C18 columns.
  • Achieves excellent efficiency and performance
  • Resolves complicated mixtures and increases throughput
  • High level of retention and stability
  • Enables faster sample preparation with high organic solvent concentration mobile phases
  • Increases MS response and sensitivity
  • Reduces A, B, and C terms of the van Deemter equation for improved performance
  • Manufactured to high standards under strict and controlled environments