Learn More
Waters Corp CORTECS HILIC Column, 90 Å, 2.7 μm, 2.1 × 100 mm
Supplier:  Waters Corp 176003282
Encompass_Preferred
These columns utilize solid-core particle technology to deliver high column efficiencies when paired with low-dispersive UPLC instrumentation. They offer seven distinct chemistries, including HILIC, providing versatility for rapidly separating a diverse array of compounds. Compared to fully porous particles of a similar size, solid-core columns produce sharper and narrower peaks, contributing to enhanced resolution, speed, and sensitivity. Additionally, these columns are available with 2.7 μm particles, suitable for use with HPLC and UHPLC systems.
  • Utilizes solid-core particle technology for high column efficiencies.
  • Offers seven distinct chemistries, including HILIC, for versatile compound separation.
  • Produces sharper and narrower peaks for enhanced resolution, speed, and sensitivity.
  • Available with 2.7 μm particles, suitable for use with HPLC and UHPLC systems.
Catalog No. 50-239-1033