Learn More
Waters Corp CORTECS HILIC Column, 90Å, 2.7 μm, 4.6 mm X 30 mm
Supplier:  Waters Corp 176003317
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle that are designed to retain extremely polar analytes. They offer orthogonal selectivity versus C18 columns.
  • High-efficiency columns based on a solid-core particle
  • Designed to retain extremely polar analytes
  • Offers orthogonal selectivity compared to C18 columns
Catalog No. 50-239-0621