Learn More
Waters Corp CORTECS UPLC Column Method Validation Kit, HILIC, 2.7 μm, 3.0 × 50 mm
Catalog No. 502390462 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS UPLC Column Method Validation Kit is designed for HILIC chemistry applications. It features a particle size of 2.7 μm and dimensions of 3.0 × 50 mm. Each kit is supplied with three columns, each from a different batch of material, to facilitate method validation.
  • HILIC chemistry for polar compound analysis
  • Particle size of 2.7 μm for efficient separations
  • Dimensions of 3.0 × 50 mm
  • Kit includes three columns
  • Columns sourced from three different material batches for robust method validation
Catalog No. 50-239-0462 Supplier Waters Corp Supplier No. 186007430
This CORTECS UPLC Column Method Validation Kit is designed for HILIC chemistry applications. It features a particle size of 2.7 μm and dimensions of 3.0 × 50 mm. Each kit is supplied with three columns, each from a different batch of material, to facilitate method validation.
  • HILIC chemistry for polar compound analysis
  • Particle size of 2.7 μm for efficient separations
  • Dimensions of 3.0 × 50 mm
  • Kit includes three columns
  • Columns sourced from three different material batches for robust method validation