Learn More
Waters Corp CORTECS UPLC Columns Method Validation Kit, HILIC, 1.6 μm, 2.1 x 75 mm
Catalog No. 502391122 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC Columns Method Validation Kit is designed for HILIC applications. It features 1.6 μm particle size columns with dimensions of 2.1 × 75 mm. This kit is suitable for method validation processes in demanding laboratory environments.
  • Designed for HILIC applications
  • Features 1.6 μm particle size columns
  • Dimensions of 2.1 × 75 mm
  • Kit contains three columns from three batches of material
Catalog No. 50-239-1122 Supplier Waters Corp Supplier No. 186007168
The CORTECS UPLC Columns Method Validation Kit is designed for HILIC applications. It features 1.6 μm particle size columns with dimensions of 2.1 × 75 mm. This kit is suitable for method validation processes in demanding laboratory environments.
  • Designed for HILIC applications
  • Features 1.6 μm particle size columns
  • Dimensions of 2.1 × 75 mm
  • Kit contains three columns from three batches of material