Learn More
Waters Corp CORTECS HILIC Method Validation Kit, 90Å, 1.6 μm, 3 mm × 75 mm
Catalog No. 502391142 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS HILIC Method Validation Kit is designed for the validation of hydrophilic interaction liquid chromatography methods. It features high-efficiency columns based on a solid-core particle, ideal for retaining extremely polar analytes and offering orthogonal selectivity compared to C18 columns. Each kit contains 3 columns with 90Å pore size, 1.6 μm particle size, and dimensions of 3 mm × 75 mm.
  • High-efficiency columns based on a solid-core particle.
  • Designed to retain extremely polar analytes.
  • Offers orthogonal selectivity vs. C18 columns.
  • Kit contains 3 columns.
Catalog No. 50-239-1142 Supplier Waters Corp Supplier No. 186007173
The CORTECS HILIC Method Validation Kit is designed for the validation of hydrophilic interaction liquid chromatography methods. It features high-efficiency columns based on a solid-core particle, ideal for retaining extremely polar analytes and offering orthogonal selectivity compared to C18 columns. Each kit contains 3 columns with 90Å pore size, 1.6 μm particle size, and dimensions of 3 mm × 75 mm.
  • High-efficiency columns based on a solid-core particle.
  • Designed to retain extremely polar analytes.
  • Offers orthogonal selectivity vs. C18 columns.
  • Kit contains 3 columns.