Learn More
Waters Corp CORTECS HILIC Method Validation Kit, 90Å, 2.7 μm, 2.1 mm × 30 mm
Catalog No. 502392008 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle designed to retain extremely polar analytes. They offer orthogonal selectivity compared to C18 columns.
  • High-efficiency columns
  • Solid-core particle
  • Designed to retain extremely polar analytes
  • Offers orthogonal selectivity versus C18 columns
Catalog No. 50-239-2008 Supplier Waters Corp Supplier No. 186007424
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle designed to retain extremely polar analytes. They offer orthogonal selectivity compared to C18 columns.
  • High-efficiency columns
  • Solid-core particle
  • Designed to retain extremely polar analytes
  • Offers orthogonal selectivity versus C18 columns