Learn More
Waters Corp UPLC HILIC Column Method Validation Kit, 1.6 μm, 90 Å, 2.1 × 150 mm
Catalog No. 502392565 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC HILIC Column Method Validation Kit is designed to evaluate the quality, reliability, and consistency of an analytical method. It includes three columns from three different batches of material to ensure batch-to-batch and column-to-column reproducibility.
  • Optimized for UltraPerformance Liquid Chromatography (UPLC).
  • Utilizes sub-2 μm solid-core particle technology.
  • Ensures reproducibility of chromatographic performance.
  • Kit contains three columns from three batches of material.
Catalog No. 50-239-2565 Supplier Waters Corp Supplier No. 186007170
The CORTECS UPLC HILIC Column Method Validation Kit is designed to evaluate the quality, reliability, and consistency of an analytical method. It includes three columns from three different batches of material to ensure batch-to-batch and column-to-column reproducibility.
  • Optimized for UltraPerformance Liquid Chromatography (UPLC).
  • Utilizes sub-2 μm solid-core particle technology.
  • Ensures reproducibility of chromatographic performance.
  • Kit contains three columns from three batches of material.