Learn More
Waters Corp CORTECS HILIC Method Validation Kit, 90Å, 2.7 μm, 4.6 mm × 50 mm
Catalog No. 502390837 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HILIC columns are high-efficiency columns built on a solid-core particle designed to retain extremely polar analytes. They offer orthogonal selectivity compared to C18 columns.
  • High-efficiency columns
  • Based on solid-core particles
  • Designed to retain extremely polar analytes
  • Offers orthogonal selectivity vs. C18 columns
Catalog No. 50-239-0837 Supplier Waters Corp Supplier No. 186007435
These HILIC columns are high-efficiency columns built on a solid-core particle designed to retain extremely polar analytes. They offer orthogonal selectivity compared to C18 columns.
  • High-efficiency columns
  • Based on solid-core particles
  • Designed to retain extremely polar analytes
  • Offers orthogonal selectivity vs. C18 columns