Learn More
Waters Corp Method Validation Kit, HILIC, 90Å/2.7 μm, 2.1 mm × 75 mm
Catalog No. 502390936 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle designed to retain extremely polar analytes. They offer orthogonal selectivity compared to C18 columns. Waters has removed paper COAs and eCords from select columns for easy access with less waste; all column documents are now available at www.waters.com/coa.
  • Hydrophilic interaction (HILIC) separation mode
  • Silica particle substrate
  • pH range of 1 to 5
  • Maximum pressure of 18000 psi (1240 bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 2.7 μm particle size
  • Parker-style endfitting type
  • 90 Å pore size
  • Method validation kit format
  • USP classification L3
  • 2.1 mm inner diameter
  • 75 mm length
  • 3 units per package
Catalog No. 50-239-0936 Supplier Waters Corp Supplier No. 186007426
CORTECS HILIC Columns are high-efficiency columns based on a solid-core particle designed to retain extremely polar analytes. They offer orthogonal selectivity compared to C18 columns. Waters has removed paper COAs and eCords from select columns for easy access with less waste; all column documents are now available at www.waters.com/coa.
  • Hydrophilic interaction (HILIC) separation mode
  • Silica particle substrate
  • pH range of 1 to 5
  • Maximum pressure of 18000 psi (1240 bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 2.7 μm particle size
  • Parker-style endfitting type
  • 90 Å pore size
  • Method validation kit format
  • USP classification L3
  • 2.1 mm inner diameter
  • 75 mm length
  • 3 units per package