Learn More
Waters Corp CORTECS HILIC Method Validation Kit, 1.6 μm, 2.1 x 30 mm, 3/pk
Catalog No. 502391116 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS HILIC Method Validation Kit is engineered to evaluate the quality, reliability, and consistency of analytical methods. It ensures batch-to-batch and column-to-column reproducibility, providing confidence in the continued use of methods. This kit contains three columns from three batches of material, with a particle size of 1.6 μm and dimensions of 2.1 × 30 mm.
  • Designed to evaluate quality, reliability, and consistency of an analytical method.
  • Includes three batches of chromatographic media derived from different base particles.
  • Ensures batch-to-batch and column-to-column reproducibility.
Catalog No. 50-239-1116 Supplier Waters Corp Supplier No. 186007166
This CORTECS HILIC Method Validation Kit is engineered to evaluate the quality, reliability, and consistency of analytical methods. It ensures batch-to-batch and column-to-column reproducibility, providing confidence in the continued use of methods. This kit contains three columns from three batches of material, with a particle size of 1.6 μm and dimensions of 2.1 × 30 mm.
  • Designed to evaluate quality, reliability, and consistency of an analytical method.
  • Includes three batches of chromatographic media derived from different base particles.
  • Ensures batch-to-batch and column-to-column reproducibility.