Learn More
Waters Corp WATERS CORPORATION CORTECS UPLC Method Validation Kit, HILIC, 2.7 μm, 2.1 x 100 mm
Catalog No. 502038013 Shop All Waters Corporation Products
Encompass_Preferred
This Method Validation Kit features CORTECS UPLC Columns designed for HILIC chemistry. It includes columns with a 2.7 μm particle size and dimensions of 2.1 x 100 mm, supplied as a 3-pack.
  • Ensures batch-to-batch and column-to-column reproducibility for reliable analytical methods.
  • Includes three batches of chromatographic media to evaluate method quality, reliability, and consistency.
  • Designed for HILIC chemistry.
  • Contains columns with 2.7 μm particle size.
  • Columns have dimensions of 2.1 x 100 mm.
  • Kit includes three columns.
Catalog No. 50-203-8013 Supplier Waters Corp Supplier No. 186007427
This Method Validation Kit features CORTECS UPLC Columns designed for HILIC chemistry. It includes columns with a 2.7 μm particle size and dimensions of 2.1 x 100 mm, supplied as a 3-pack.
  • Ensures batch-to-batch and column-to-column reproducibility for reliable analytical methods.
  • Includes three batches of chromatographic media to evaluate method quality, reliability, and consistency.
  • Designed for HILIC chemistry.
  • Contains columns with 2.7 μm particle size.
  • Columns have dimensions of 2.1 x 100 mm.
  • Kit includes three columns.