Learn More
Waters Corp CORTECS VanGuard Cartridge, HILIC, 2.7 μm, 3.9 × 5 mm
Catalog No. 502392961 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridge columns extend the lifespan of any UHPLC column. The original separation performance can be restored by simply replacing the cartridge, which is more cost-effective than replacing the entire analytical column.
  • Removes microparticles and chemical contaminants
  • Maintains UHPLC separation efficiency
  • Provides cost-effective column protection
  • Reusable holder connects to any column inlet as the ferrule is not fixed before the initial connection
Catalog No. 50-239-2961 Supplier Waters Corp Supplier No. 186007690
VanGuard Cartridge columns extend the lifespan of any UHPLC column. The original separation performance can be restored by simply replacing the cartridge, which is more cost-effective than replacing the entire analytical column.
  • Removes microparticles and chemical contaminants
  • Maintains UHPLC separation efficiency
  • Provides cost-effective column protection
  • Reusable holder connects to any column inlet as the ferrule is not fixed before the initial connection