Learn More
Waters Corp CORTECS VanGuard Cartridge, HILIC, 90Å, 2.7 μm, 2.1 × 5 mm
Catalog No. 502390959 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS HILIC VanGuard Cartridge protects analytical columns from particulate and chemical contamination, extending their lifetime and performance. Optimized for 2.1 mm and 3.0 mm I.D. HILIC columns with 2.7 μm sorbent, it offers high efficiency and orthogonal selectivity for retaining polar analytes and increasing throughput.
  • Extends analytical column lifetime by removing contaminants
  • Optimized for 2.1 mm and 3.0 mm I.D. columns with 2.7 μm sorbent
  • Provides high efficiency and orthogonal selectivity for polar analytes
  • Enables faster separations and increased throughput
  • Facilitates direct operation with SPE eluates
  • Improves MS response and sensitivity
Catalog No. 50-239-0959 Supplier Waters Corp Supplier No. 186007688
This CORTECS HILIC VanGuard Cartridge protects analytical columns from particulate and chemical contamination, extending their lifetime and performance. Optimized for 2.1 mm and 3.0 mm I.D. HILIC columns with 2.7 μm sorbent, it offers high efficiency and orthogonal selectivity for retaining polar analytes and increasing throughput.
  • Extends analytical column lifetime by removing contaminants
  • Optimized for 2.1 mm and 3.0 mm I.D. columns with 2.7 μm sorbent
  • Provides high efficiency and orthogonal selectivity for polar analytes
  • Enables faster separations and increased throughput
  • Facilitates direct operation with SPE eluates
  • Improves MS response and sensitivity