Learn More
Waters Corp Waters CORTECS VanGuard Pre-column, HILIC, 90Å, 1.6 μm, 2.1 mm x 5 mm
Catalog No. 502037847 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-efficiency pre-column protects analytical columns and enhances HILIC column performance for polar analytes.
  • Features HILIC separation mode with silica particle substrate.
  • pH range 1 to 5.
  • Max pressure 18000 psi (1240 Bar).
  • Not endcapped; high silanol activity.
  • Spherical solid core particles: 1.6 μm size, 90 Å pore, 100 surface area.
  • Direct-Connect endfitting.
  • UPLC and UHPLC system compatible.
  • L3 USP classification.
  • Dimensions: 2.1 mm inner diameter, 5 mm length.
  • Protects and extends analytical column life.
  • Improves selectivity, sensitivity, and analysis speed.
  • Suitable for biological fluid purification and temperature control.
  • Manufactured in cGMP, ISO 9001 certified plant; chromatographically tested with ultra-pure reagents.
Catalog No. 50-203-7847 Supplier Waters Corp Supplier No. 186007124
This high-efficiency pre-column protects analytical columns and enhances HILIC column performance for polar analytes.
  • Features HILIC separation mode with silica particle substrate.
  • pH range 1 to 5.
  • Max pressure 18000 psi (1240 Bar).
  • Not endcapped; high silanol activity.
  • Spherical solid core particles: 1.6 μm size, 90 Å pore, 100 surface area.
  • Direct-Connect endfitting.
  • UPLC and UHPLC system compatible.
  • L3 USP classification.
  • Dimensions: 2.1 mm inner diameter, 5 mm length.
  • Protects and extends analytical column life.
  • Improves selectivity, sensitivity, and analysis speed.
  • Suitable for biological fluid purification and temperature control.
  • Manufactured in cGMP, ISO 9001 certified plant; chromatographically tested with ultra-pure reagents.