Learn More
Waters Corp Column, Phenyl, 90Å, 1.6 μm, 3 mm x 75 mm
Catalog No. 502390703 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns offer complementary selectivity to C18 stationary phases. Their particle morphology contributes to high efficiency and lower backpressures compared to fully porous particles of similar size. They provide ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Offer complementary selectivity to C18 stationary phases
  • Contribute to high efficiency
  • Provide lower backpressures compared to fully porous particles of similar size
  • Ensure ultra-low column MS bleed
  • Support long lifetimes
  • Deliver excellent peak shape
Catalog No. 50-239-0703 Supplier Waters Corp Supplier No. 176003826
Add to Cart
Add to Cart
These columns offer complementary selectivity to C18 stationary phases. Their particle morphology contributes to high efficiency and lower backpressures compared to fully porous particles of similar size. They provide ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Offer complementary selectivity to C18 stationary phases
  • Contribute to high efficiency
  • Provide lower backpressures compared to fully porous particles of similar size
  • Ensure ultra-low column MS bleed
  • Support long lifetimes
  • Deliver excellent peak shape