Learn More
Waters Corp CORTECS Phenyl Column, 90 Å, 1.6 μm, 3 mm x 50 mm
Catalog No. 502390843 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Phenyl columns offer complementary selectivity to C18 stationary phases. The particle morphology ensures high efficiency and reduced backpressures compared to fully porous particles of similar size. These columns provide ultra-low column MS bleed, extended lifetimes, and excellent peak shape.
  • Provides complimentary selectivity to C18 stationary phases.
  • Offers high efficiency and lower backpressures.
  • Ensures ultra-low column MS bleed.
  • Delivers long lifetimes.
  • Achieves excellent peak shape.
Catalog No. 50-239-0843 Supplier Waters Corp Supplier No. 176003825
CORTECS Phenyl columns offer complementary selectivity to C18 stationary phases. The particle morphology ensures high efficiency and reduced backpressures compared to fully porous particles of similar size. These columns provide ultra-low column MS bleed, extended lifetimes, and excellent peak shape.
  • Provides complimentary selectivity to C18 stationary phases.
  • Offers high efficiency and lower backpressures.
  • Ensures ultra-low column MS bleed.
  • Delivers long lifetimes.
  • Achieves excellent peak shape.