Learn More
Waters Corp CORTECS Phenyl Column, Phenyl, 90Å, 2.7 μm, 2.1 mm X 75 mm
Catalog No. 502391346 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns offer complementary selectivity to C18 stationary phases. Their particle morphology provides high efficiency and lower backpressures compared to fully porous particles of similar size, delivering ultra-low MS bleed, long lifetimes, and excellent peak shape.
  • Offers complementary selectivity to C18 stationary phases.
  • Provides high efficiency.
  • Delivers lower backpressures compared to fully porous particles of similar size.
  • Ensures ultra-low column MS bleed.
  • Features long lifetimes.
  • Promotes excellent peak shape.
Catalog No. 50-239-1346 Supplier Waters Corp Supplier No. 186008320
These columns offer complementary selectivity to C18 stationary phases. Their particle morphology provides high efficiency and lower backpressures compared to fully porous particles of similar size, delivering ultra-low MS bleed, long lifetimes, and excellent peak shape.
  • Offers complementary selectivity to C18 stationary phases.
  • Provides high efficiency.
  • Delivers lower backpressures compared to fully porous particles of similar size.
  • Ensures ultra-low column MS bleed.
  • Features long lifetimes.
  • Promotes excellent peak shape.