Learn More
Waters Corp CORTECS Phenyl Column, 90Å, 2.7 μm, 4.6 mm × 75 mm
Catalog No. 502391678 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS 2.7 μm Columns are based on solid-core particle technology, allowing for method transfer between UPLC, UHPLC, and HPLC platforms. They are manufactured in cGMP, ISO 9001:2000 certified facilities, with each column individually tested.
  • Manufactured in cGMP, ISO 9001:2000 certified facilities, ensuring high quality and reproducible products.
  • eCord Intelligent Chip technology available on 2.1 mm and 3.0 mm column dimensions, providing paperless tracking history of column performance and usage.
  • Designed to operate on any HPLC, UHPLC, or UPLC system.
  • Solid-core particle technology enables seamless method transfer across different chromatography platforms.
  • VanGuard™ Cartridge columns offer a universal solution to extend the lifetime of all HPLC, UPLC, and UHPLC columns.
Catalog No. 50-239-1678 Supplier Waters Corp Supplier No. 176003801
CORTECS 2.7 μm Columns are based on solid-core particle technology, allowing for method transfer between UPLC, UHPLC, and HPLC platforms. They are manufactured in cGMP, ISO 9001:2000 certified facilities, with each column individually tested.
  • Manufactured in cGMP, ISO 9001:2000 certified facilities, ensuring high quality and reproducible products.
  • eCord Intelligent Chip technology available on 2.1 mm and 3.0 mm column dimensions, providing paperless tracking history of column performance and usage.
  • Designed to operate on any HPLC, UHPLC, or UPLC system.
  • Solid-core particle technology enables seamless method transfer across different chromatography platforms.
  • VanGuard™ Cartridge columns offer a universal solution to extend the lifetime of all HPLC, UPLC, and UHPLC columns.