Learn More
Waters Corp CORTECS Phenyl Column, 90 Å, 2.7 μm, 2.1 mm × 75 mm
Catalog No. 502391879 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Phenyl columns offer complementary selectivity to C18 stationary phases. Their particle morphology provides high efficiency and lower backpressures compared to fully porous particles of similar size.
  • High efficiency
  • Lower backpressures compared to fully porous particles of similar size
  • Ultra-low column MS bleed
  • Long lifetimes
  • Excellent peak shape
Catalog No. 50-239-1879 Supplier Waters Corp Supplier No. 176003791
Add to Cart
Add to Cart
CORTECS Phenyl columns offer complementary selectivity to C18 stationary phases. Their particle morphology provides high efficiency and lower backpressures compared to fully porous particles of similar size.
  • High efficiency
  • Lower backpressures compared to fully porous particles of similar size
  • Ultra-low column MS bleed
  • Long lifetimes
  • Excellent peak shape