Learn More
Waters Corp Column, Phenyl, 2.7 μm, 4.6 mm Inner Diameter, 50 mm
Catalog No. 502392133 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column provides complimentary selectivity to C18 stationary phases. Its particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size. It provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Provides complimentary selectivity to C18 stationary phases.
  • Particle morphology offers high efficiency and lower backpressures compared to fully porous particles of similar particle size.
  • Offers ultra-low column MS bleed.
  • Ensures long lifetimes.
  • Delivers excellent peak shape.
Catalog No. 50-239-2133 Supplier Waters Corp Supplier No. 186008339
This column provides complimentary selectivity to C18 stationary phases. Its particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size. It provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Provides complimentary selectivity to C18 stationary phases.
  • Particle morphology offers high efficiency and lower backpressures compared to fully porous particles of similar particle size.
  • Offers ultra-low column MS bleed.
  • Ensures long lifetimes.
  • Delivers excellent peak shape.