Learn More
Waters Corp CORTECS Phenyl Method Validation Kit, 90Å, 1.6 μm, 3 mm X 75 mm, 3/pk
Catalog No. 502390735 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These method validation kits include columns from three different batches. The columns offer complementary selectivity to C18 stationary phases. Their particle morphology ensures high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Complimentary selectivity to C18 stationary phases
  • High efficiency and lower backpressures
  • Ultra-low column MS bleed
  • Long lifetimes
  • Excellent peak shape
Catalog No. 50-239-0735 Supplier Waters Corp Supplier No. 186008395
These method validation kits include columns from three different batches. The columns offer complementary selectivity to C18 stationary phases. Their particle morphology ensures high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Complimentary selectivity to C18 stationary phases
  • High efficiency and lower backpressures
  • Ultra-low column MS bleed
  • Long lifetimes
  • Excellent peak shape