Learn More
Waters Corp CORTECS Phenyl Method Validation Kit, 90Å, 1.6 μm, 2.1 x 30 mm
Catalog No. 502391495 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS Phenyl Method Validation Kit is designed for comprehensive method validation applications. This kit provides columns with complementary selectivity to C18 stationary phases, making it suitable for separating a variety of compounds.
  • Columns of three different batches
  • Complementary selectivity to C18 stationary phases
  • Ideal for method validation applications
  • Features 90Å pore size and 1.6 μm particle size
  • Dimensions are 2.1 mm ID x 30 mm Length
Catalog No. 50-239-1495 Supplier Waters Corp Supplier No. 186008383
The CORTECS Phenyl Method Validation Kit is designed for comprehensive method validation applications. This kit provides columns with complementary selectivity to C18 stationary phases, making it suitable for separating a variety of compounds.
  • Columns of three different batches
  • Complementary selectivity to C18 stationary phases
  • Ideal for method validation applications
  • Features 90Å pore size and 1.6 μm particle size
  • Dimensions are 2.1 mm ID x 30 mm Length