Learn More
Waters Corp CORTECS Method Validation Kit, Phenyl, 90Å, 1.6 μm
Catalog No. 502391934 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Phenyl method validation kits provide columns of three different batches. These columns offer complimentary selectivity to C18 stationary phases. Their particle morphology ensures high efficiency and lower back pressures compared to fully porous particles of similar size, while also delivering ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Provides columns of three different batches for method validation.
  • Offers complimentary selectivity to C18 stationary phases.
  • Particle morphology ensures high efficiency and lower back pressures.
  • Delivers ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Manufactured in a cGMP, ISO 9001 accredited facility.
  • Designed exclusively for the ACQUITY UPLC System.
  • Each batch chromatographically tested for consistent performance.
Catalog No. 50-239-1934 Supplier Waters Corp Supplier No. 186008384
CORTECS Phenyl method validation kits provide columns of three different batches. These columns offer complimentary selectivity to C18 stationary phases. Their particle morphology ensures high efficiency and lower back pressures compared to fully porous particles of similar size, while also delivering ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Provides columns of three different batches for method validation.
  • Offers complimentary selectivity to C18 stationary phases.
  • Particle morphology ensures high efficiency and lower back pressures.
  • Delivers ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Manufactured in a cGMP, ISO 9001 accredited facility.
  • Designed exclusively for the ACQUITY UPLC System.
  • Each batch chromatographically tested for consistent performance.