Learn More
Waters Corp CORTECS Phenyl Method Validation Kit, 90 Å, 1.6 μm
Catalog No. 502392250 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Phenyl method validation kits offer columns from three distinct batches, providing complimentary selectivity when compared to C18 stationary phases. The particle morphology contributes to high efficiency and reduced backpressures, especially when contrasted with fully porous particles of similar size.
  • Ensures ultra-low MS bleed
  • Provides extended lifetimes
  • Delivers excellent peak shape
  • Enhances reliability and performance of chromatographic analyses
Catalog No. 50-239-2250 Supplier Waters Corp Supplier No. 186008397
CORTECS Phenyl method validation kits offer columns from three distinct batches, providing complimentary selectivity when compared to C18 stationary phases. The particle morphology contributes to high efficiency and reduced backpressures, especially when contrasted with fully porous particles of similar size.
  • Ensures ultra-low MS bleed
  • Provides extended lifetimes
  • Delivers excellent peak shape
  • Enhances reliability and performance of chromatographic analyses