Learn More
Waters Corp CORTECS Method Validation Kit, Phenyl, 1.6 μm, 2.1 mm x 100 mm
Catalog No. 502392432 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Phenyl method validation kits include columns from three different batches, offering complementary selectivity to C18 stationary phases. Their particle morphology provides high efficiency and lower backpressures compared to fully porous particles of a similar size. These kits also ensure ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Includes columns from three different batches.
  • Offers complementary selectivity to C18 stationary phases.
  • Particle morphology provides high efficiency and reduced backpressures.
  • Ensures ultra-low column MS bleed, extended lifetimes, and excellent peak shape.
  • Features Phenyl chemistry and reversed phase separation mode.
  • Withstands a maximum pressure of 18000 psi (1240 Bar).
  • Compatible with UPLC and UHPLC systems.
  • Dimensions: 2.1 mm inner diameter, 100 mm length.
  • Includes 3 units per package.
Catalog No. 50-239-2432 Supplier Waters Corp Supplier No. 186008386
CORTECS Phenyl method validation kits include columns from three different batches, offering complementary selectivity to C18 stationary phases. Their particle morphology provides high efficiency and lower backpressures compared to fully porous particles of a similar size. These kits also ensure ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Includes columns from three different batches.
  • Offers complementary selectivity to C18 stationary phases.
  • Particle morphology provides high efficiency and reduced backpressures.
  • Ensures ultra-low column MS bleed, extended lifetimes, and excellent peak shape.
  • Features Phenyl chemistry and reversed phase separation mode.
  • Withstands a maximum pressure of 18000 psi (1240 Bar).
  • Compatible with UPLC and UHPLC systems.
  • Dimensions: 2.1 mm inner diameter, 100 mm length.
  • Includes 3 units per package.