Learn More
Waters Corp Method Validation Kit, CORTECS Phenyl, 90Å, 2.7 μm
Catalog No. 502391068 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a Method Validation Kit designed for chromatography applications. It includes CORTECS Phenyl columns with specific pore size and particle size, suitable for method development and validation.
  • Includes CORTECS Phenyl columns.
  • Features a pore size of 90 Å.
  • Designed with a particle size of 2.7 μm.
  • Each column measures 4.6 mm ID by 150 mm in length.
  • Supplied as a 3-pack kit.
  • Complies with USP L11 specification.
Catalog No. 50-239-1068 Supplier Waters Corp Supplier No. 186008347
This product is a Method Validation Kit designed for chromatography applications. It includes CORTECS Phenyl columns with specific pore size and particle size, suitable for method development and validation.
  • Includes CORTECS Phenyl columns.
  • Features a pore size of 90 Å.
  • Designed with a particle size of 2.7 μm.
  • Each column measures 4.6 mm ID by 150 mm in length.
  • Supplied as a 3-pack kit.
  • Complies with USP L11 specification.