Learn More
Waters Corp CORTECS Phenyl Method Validation Kit, 90Å, 2.7 μm, 3 x 50 mm, 3/pk
Catalog No. 502391070 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Phenyl method validation kits offer columns from three distinct batches. These columns provide complementary selectivity to C18 stationary phases. Their particle morphology delivers high efficiency and lower backpressures compared to fully porous particles of similar size. The columns also ensure ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Provides columns of three different batches for method validation.
  • Offers complementary selectivity to C18 stationary phases.
  • High efficiency and lower backpressures due to particle morphology.
  • Ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.
Catalog No. 50-239-1070 Supplier Waters Corp Supplier No. 186008334
CORTECS Phenyl method validation kits offer columns from three distinct batches. These columns provide complementary selectivity to C18 stationary phases. Their particle morphology delivers high efficiency and lower backpressures compared to fully porous particles of similar size. The columns also ensure ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Provides columns of three different batches for method validation.
  • Offers complementary selectivity to C18 stationary phases.
  • High efficiency and lower backpressures due to particle morphology.
  • Ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.