Learn More
Waters Corp CORTECS Phenyl Method Validation Kit, 90Å, 2.7μm, 3.0 x 150 mm
Catalog No. 502391074 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit features CORTECS Phenyl phase material with a particle size of 2.7 μm and a pore size of 90 Å. The columns have an inner diameter of 3.0 mm and a length of 150 mm, making them suitable for method validation applications.
  • Columns have an inner diameter of 3.0 mm.
  • Column length is 150 mm.
  • Features a particle size of 2.7 μm.
  • Pore size is 90 Å.
  • Utilizes CORTECS Phenyl phase material.
  • Designed for method validation.
  • Conforms to USP L11.
Catalog No. 50-239-1074 Supplier Waters Corp Supplier No. 186008337
This Method Validation Kit features CORTECS Phenyl phase material with a particle size of 2.7 μm and a pore size of 90 Å. The columns have an inner diameter of 3.0 mm and a length of 150 mm, making them suitable for method validation applications.
  • Columns have an inner diameter of 3.0 mm.
  • Column length is 150 mm.
  • Features a particle size of 2.7 μm.
  • Pore size is 90 Å.
  • Utilizes CORTECS Phenyl phase material.
  • Designed for method validation.
  • Conforms to USP L11.