Learn More
Waters Corp CORTECS Phenyl Method Validation Kit, 90Å, 1.6 μm, 2.1 mm X 75 mm
Catalog No. 502390924 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is a CORTECS Phenyl Method Validation Kit, featuring a pore size of 90Å, a particle size of 1.6 μm, and column dimensions of 2.1 mm X 75 mm. It includes 3 columns, making it suitable for method validation applications.
  • Pore size of 90Å for effective separation
  • Particle size of 1.6 μm ensures high resolution
  • Column dimensions of 2.1 mm X 75 mm for analytical separations
  • Kit includes 3 columns for comprehensive method validation
Catalog No. 50-239-0924 Supplier Waters Corp Supplier No. 186008385
This kit is a CORTECS Phenyl Method Validation Kit, featuring a pore size of 90Å, a particle size of 1.6 μm, and column dimensions of 2.1 mm X 75 mm. It includes 3 columns, making it suitable for method validation applications.
  • Pore size of 90Å for effective separation
  • Particle size of 1.6 μm ensures high resolution
  • Column dimensions of 2.1 mm X 75 mm for analytical separations
  • Kit includes 3 columns for comprehensive method validation