Learn More
Waters Corp WATERS CORPORATION CORTECS Phenyl Column, 90Å, 1.6 μm, 2.1x150 mm
Catalog No. 502038559 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Phenyl method validation kits provide columns of three different batches. CORTECS Phenyl columns offer complementary selectivity to C18 stationary phases. The particle morphology delivers high efficiency and lower backpressures compared to fully porous particles of similar particle size. CORTECS Phenyl provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Method validation kits provide columns of three different batches.
  • Columns offer complementary selectivity to C18 stationary phases.
  • The particle morphology provides high efficiency and lower backpressures compared to fully porous particles of similar particle size.
  • Ensures ultra-low column MS bleed, long lifetimes, and excellent peak shape.
Catalog No. 50-203-8559 Supplier Waters Corp Supplier No. 186008387
CORTECS Phenyl method validation kits provide columns of three different batches. CORTECS Phenyl columns offer complementary selectivity to C18 stationary phases. The particle morphology delivers high efficiency and lower backpressures compared to fully porous particles of similar particle size. CORTECS Phenyl provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Method validation kits provide columns of three different batches.
  • Columns offer complementary selectivity to C18 stationary phases.
  • The particle morphology provides high efficiency and lower backpressures compared to fully porous particles of similar particle size.
  • Ensures ultra-low column MS bleed, long lifetimes, and excellent peak shape.