Learn More
Waters Corp WATERS CORPORATION CORTECS Premier C18 Column, MaxPeak High Performance Surfaces, 1.6 μm, 2.1 x 150 mm
Catalog No. NC2294334 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column utilizes MaxPeak High Performance Surfaces (HPS) technology to minimize analyte/surface interactions. It enhances analyte recovery, sensitivity, and reproducibility, preventing loss of metal-sensitive analytes such as lipids, organic acids, and acidic peptides.
  • Minimizes analyte/surface interactions, increasing analyte recovery, sensitivity, and reproducibility.
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes, and enhances recovery.
  • Enables simpler mobile phases without complex additives, saving time in method development.
  • Reduces risk and increases confidence in data and decision making.
  • Utilizes solid-core technology for maximum efficiency, increased sample throughput, and lower column backpressure.
This item is not returnable. View return policy
Catalog No. NC2294334 Supplier Waters Corp Supplier No. 186009513
This column utilizes MaxPeak High Performance Surfaces (HPS) technology to minimize analyte/surface interactions. It enhances analyte recovery, sensitivity, and reproducibility, preventing loss of metal-sensitive analytes such as lipids, organic acids, and acidic peptides.
  • Minimizes analyte/surface interactions, increasing analyte recovery, sensitivity, and reproducibility.
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes, and enhances recovery.
  • Enables simpler mobile phases without complex additives, saving time in method development.
  • Reduces risk and increases confidence in data and decision making.
  • Utilizes solid-core technology for maximum efficiency, increased sample throughput, and lower column backpressure.