Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp CORTECS Premier C18 Column, 1.6 µm, 2.1 mm x 100 mm, 1/pk;

Supplier:  Waters Corp 186009512

Encompass_Preferred

CORTECS Premier C18 Column, 1.6 µm, 2.1 mm x 100 mm, 1/pk;

Catalog No. NC2294333


May include imposed supplier surcharges.
Only null left
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.