Learn More
Waters Corp Waters Column, C18+, 2.7 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186010455
Encompass_Preferred
CORTECS Premier C18+ columns are high-efficiency C18 columns with solid-core particles, engineered for excellent peak shape for basic analytes at low pH. They incorporate MaxPeak High Performance Surface technology to reduce unwanted analyte/surface interactions, preventing poor peak shape and signal intensity loss.
  • Provides excellent peak shape for basic analytes at low pH.
  • MaxPeak High Performance Surface technology minimizes analyte interactions.
  • Reduces signal loss and poor peak shape, especially for metal-sensitive analytes.
  • Enhances analyte recovery, sensitivity, and reproducibility.
  • Shortens column conditioning and method development times.
  • Allows for simpler mobile phases.
  • Maximizes efficiency and throughput while lowering backpressure.
Catalog No. NC2294362