Learn More
Waters Corp UHPLC Column, CORTECS Premier C18+, 2.7 μm, 2.1 mm x 150 mm
Catalog No. NC2294366 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Premier C18+ columns are general-purpose, high-efficiency C18 columns designed for excellent peak shape with basic analytes.
  • General purpose, high efficiency C18 columns based on a solid-core particle
  • Designed to deliver excellent peak shape for basic analytes at low pH
  • Utilizes MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity
  • Includes VanGuard Fit column design, which protects the column from contaminants and increases column lifetime
This item is not returnable. View return policy
Catalog No. NC2294366 Supplier Waters Corp Supplier No. 186010457
Add to Cart
Add to Cart
CORTECS Premier C18+ columns are general-purpose, high-efficiency C18 columns designed for excellent peak shape with basic analytes.
  • General purpose, high efficiency C18 columns based on a solid-core particle
  • Designed to deliver excellent peak shape for basic analytes at low pH
  • Utilizes MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity
  • Includes VanGuard Fit column design, which protects the column from contaminants and increases column lifetime