Learn More
Waters Corp Waters CORTECS Premier C18 Column, 2.7 μm, 2.1 mm, 100 mm
Catalog No. NC2294354 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS Premier C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
  • All column documents are now available online, with paper certificates removed for less waste.
This item is not returnable. View return policy
Catalog No. NC2294354 Supplier Waters Corp Supplier No. 186010442
The CORTECS Premier C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
  • All column documents are now available online, with paper certificates removed for less waste.